2',3'-O-Isopropylideneadenosine 98%

Supplier: Ambeed
Warning

A177625-100G A177625-10G A177625-1G A177625-250MG A177625-25G A177625-5G A177625-500G
103845-584EA 14 USD
103845-584 76720-728 76720-730 76720-732 76720-734 77082-970 77082-972 77879-171
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine

Caution: for Research Use Only

Formula: C₁₃H₁₇N₅O₄
Melting Pt: 221…222 °C
Flash Pt: 298.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00005756
CAS Number: 362-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR