Methyl stearate 99+%

Supplier: Ambeed
Warning

A171330-100G A171330-25G A171330-500G
76720-534EA 14 USD
76720-534 76720-536 76720-538
Methyl stearate 99+%
Methyl stearate

Caution: for Research Use Only

Formula: CH₃(CH₂)₁₆CO₂CH₃
MDL Number: MFCD00009005
CAS Number: 112-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR