Ethyl indol-3-ylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-indoleacetate, Ethyl 2-(1H-indol-3-yl)acetate, Ethyl 3-Indoleacetate, IAA ethyl ester, Indole-3-acetic acid ethyl ester, Ethyl indole-3-acetate, Indol-3-ylacetic acid ethyl ester, 3-Indoleacetic acid ethyl ester

A166156-100G A166156-10G A166156-1G A166156-25G A166156-5G
76720-310EA 167 USD
76720-310 76720-312 76720-314 76720-316 76720-318
Ethyl indol-3-ylacetate 98%
Ethyl indol-3-ylacetate

Caution: for Research Use Only

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD00005635
CAS Number: 778-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR