Octyl acetate 98%

Supplier: Ambeed
Danger

A153402-25G A153402-100G A153402-500G
76719-808EA 7 USD
76719-808 77600-178 77667-164
Octyl acetate 98%
Octyl acetate

Caution: for Research Use Only

Formula: C₁₀H₂₀O₂
MDL Number: MFCD00009562
CAS Number: 112-14-1
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR