Ruthenium(III) chloride hydrate 98%

Supplier: Ambeed
Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

A151048-10G A151048-1G A151048-5G
76719-724EA 302 USD
76719-724 76719-726 76719-728
Ruthenium(III) chloride hydrate 98%
Ruthenium (III) chloride hydrate

Caution: for Research Use Only

Formula: Cl₃Ru·xH₂O
MDL Number: MFCD00149844
CAS Number: 14898-67-0
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR