Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 97%

Supplier: Ambeed
Warning

Synonyms: Kobayashi's Reagent, NaBArF24

A136880-100G A136880-10G A136880-250MG A136880-1G A136880-25G A136880-5G
103802-368EA 15 USD
103802-368 103833-350 76719-224 76719-226 76719-228 77315-286 77315-288 77315-290
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 97%
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Caution: for Research Use Only

Formula: C₃₂H₁₂BF₂₄Na
Storage Temperature: Ambient
MDL Number: MFCD00043323
CAS Number: 79060-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR