MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 99%

Supplier: Ambeed
Warning

A136758-1000G A136758-100G A136758-25G A136758-500G
76719-214EA 77 USD
76719-214 76719-216 76719-218 76719-220 76719-222
MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 99%
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)

Caution: for Research Use Only

Formula: C₇H₁₅NO₅S
MDL Number: MFCD00009629
CAS Number: 68399-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR