4-Carboxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

A132897-100G A132897-10G A132897-500G A132897-1G A132897-25G A132897-5G
103801-800EA 8 USD
103801-800 103832-722 76719-014 76719-016 76719-018 77314-382 77314-384 77314-386
4-Carboxyphenylboronic acid pinacol ester 98%
4-Carboxyphenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₃H₁₇BO₄
Boiling Pt: 379.7 °C (760 mm Hg)
Melting Pt: 228…231 °C
Storage Temperature: Ambient
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR