MS-275 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridin-3-ylmethyl 4-((2-aminophenyl)carbamoyl)benzylcarbamate, Entinostat
A122285-1MG
A122285-50MG
A122285-10MG
A122285-100MG
A122285-250MG
A122285-5G
76718-590EA
9
USD
76718-590
76718-592
77599-762
77599-760
77666-752
77878-902
MS-275 98%
MS-275
Caution: for Research Use Only
Formula:
C₂₁H₂₀N₄O₃ |
MDL Number:
MFCD08272435 CAS Number: 209783-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...