MS-275 98%

Supplier: Ambeed
Warning

Synonyms: Pyridin-3-ylmethyl 4-((2-aminophenyl)carbamoyl)benzylcarbamate, Entinostat

A122285-1MG A122285-50MG A122285-10MG A122285-100MG A122285-250MG A122285-5G
76718-590EA 9 USD
76718-590 76718-592 77599-762 77599-760 77666-752 77878-902
MS-275 98%
MS-275

Caution: for Research Use Only

Formula: C₂₁H₂₀N₄O₃
MDL Number: MFCD08272435
CAS Number: 209783-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR