Sodium dodecyl sulfate (SDS) 98%

Supplier: Ambeed
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

A120709-25G
76718-518EA 11 USD
76718-518
Sodium dodecyl sulfate (SDS) 98%
Sodium dodecyl sulphate (SDS)

Caution: for Research Use Only

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 1325
ADR: 4.1 Ⅲ
Merck Index: 14,08636

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR