n-Dodecyl-β-D-maltoside 97%

Supplier: Ambeed
Warning

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

A112981-10G A112981-250MG A112981-5G A112981-25G A112981-1G
76718-186EA 176 USD
76718-186 76718-188 76718-190 77549-190 77570-714
n-Dodecyl-β-D-maltoside 97%
n-Dodecyl-β-D-maltoside

Caution: for Research Use Only

Formula: C₂₄H₄₆O₁₁
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR