4-Bromobenzophenone 98%

Supplier: Ambeed
Warning

A105644-100G A105644-10G A105644-25G A105644-500G
103822-654EA 5 USD
103822-654 76717-792 76717-794 76717-796 76717-798
4-Bromobenzophenone 98%
4-Bromobenzophenone

Caution: for Research Use Only

Formula: C₁₃H₉BrO
Boiling Pt: 346.8 °C (760 mm Hg)
Melting Pt: 78…82 °C
Flash Pt: 81.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00000103
CAS Number: 90-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR