2-(Dimethylamino)-2-oxoacetic acid 97%

Supplier: Ambeed
Danger

A105385-100G A105385-1G A105385-25G A105385-500G A105385-5G
76717-770EA 72 USD
76717-770 76717-772 76717-774 76717-776 76717-778
2-(Dimethylamino)-2-oxoacetic acid 97%
2-(Dimethylamino)-2-oxoacetic acid

Caution: for Research Use Only

Formula: C₄H₇NO₃
MDL Number: MFCD01971023
CAS Number: 32833-96-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR