Linearity FD D-Dimer for Stago Systems, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K958M-5
76717-394EA 416.77 USD
76717-394
Linearity FD D-Dimer for Stago Systems, AUDIT Microcontrols
Controls and Calibration Verification Materials
Linearity FD D-Dimer for Stago Systems is assayed quality control material consisting of five levels of human plasma.
  • Each level contains the following analytes: D-Dimer
  • These five levels demonstrate a linear relationship to each other for their respective analytes
  • It is intended to simulate human patient plasma samples for purpose of determining linearity, calibration verification and verification of reportable range for D-Dimer.