Corepoint® Scientific General Purpose Manual Defrost Freezers (17 cu. ft.)
Supplier: Horizon Scientific
NSPF171WWW/0MHC
76717-382EA 4818.21 USD
76717-382
Corepoint® Scientific General Purpose Manual Defrost Freezers (17 cu. ft.)
Freezers Laboratory / General Purpose Freezers
These General Purpose Manual Defrost Freezers are ideal for laboratory applications.
  • Laboratory use
  • Microprocessor temperature controller
  • Spacious shelving
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
General Purpose Manual Defrost Freezers take a large step forward in performance from past general purpose units; offering a digital temperature display and alarm module coupled with a microprocessor temperature controller.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76717-382