Corepoint® Scientific Flammable Material Storage Manual Defrost Freezers (17 cu. ft.)
Supplier: Horizon Scientific
FF171WWW/0MHC
76717-362EA 5633.97 USD
76717-362
Corepoint® Scientific Flammable Material Storage Manual Defrost Freezers (17 cu. ft.)
Freezers Flammable Material Freezers
Safeguard volatile substances in these specially-designed flammable storage freezers.
  • Laboratory use
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access and microprocessor temperature control.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76717-362