AccuForge® GEM RUNNER™ Dispenser
Supplier: Accutec Blades
AGTL-7000-0000
76717-342CS 938.51 USD
76717-342
AccuForge® GEM RUNNER™ Dispenser
Blade Removers and Dispensers
GEM RUNNER™ features a reloadable single edge blade safety dispenser for cleanroom and medical manufacturing applications.
  • Holds all GEM3 and GEM2 cartridges
  • Fits 0.009" and 0.012" single edge blade GEM cartridges
  • Designed for safety and speed
  • Great for high volume uses
  • Blades not included
GEM RUNNER™ reduces the risk of injury, streamlines cutting operations, and minimizes waste. It serves up blades at the touch of a button. It’s compact and reliable, and it’s the only reloadable pop-up blade dispenser on the market.
Ordering information: Blades not included (must be ordered separately).
Packaging: 5 dispensers per case.