AccuForge® Single Edge Blades, 2 Facet
Supplier: Accutec Blades
AGBL-7032-0000 AGBL-7033-0000
76717-328CS 808.34 USD
76717-328 76717-330
AccuForge® Single Edge Blades, 2 Facet
Razors and Blades
AccuForge single edge blades feature 2-facet edges for durability. Blades are wrapped in protective paper.
  • 2-facet edge for durability
  • Re-closable clamshell for safe and convenient blade storage
  • Each clamshell contains 100 blades
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.