AccuForge® Double Edge Blades, 10 Blade Dispenser
Supplier: Accutec Blades
AGBL-6007-0000
76717-296CS 3348.27 USD
76717-296
AccuForge® Double Edge Blades, 10 Blade Dispenser
Razors and Blades
AccuForge double edge blades feature a 3-Facet edge for exacting, clean cuts. The MicroCoat® Enrichment technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. Safety blade dispenser holds 10 blades.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • 3-facet edge for increased sharpness
  • Each dispenser holds 10 blades
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.