AccuForge® Double Edge Blades, Individually Wrapped
Supplier: Accutec Blades
AGBL-6000-0000
76717-290CS 2512.43 USD
76717-290
AccuForge® Double Edge Blades, Individually Wrapped
Razors and Blades
AccuForge double edge blades feature a 3-Facet edge for exacting, clean cuts. Blades are individually wrapped for safety.
  • 3-facet blade edge for increased sharpness
  • High quality stainless steel
  • Available with MicroCoat® enrichment technology
  • Made in USA
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.