Lighthouse Locking Wall Mount Unit for Sharps Containers
Supplier: Unimed-Midwest
SWMU000609
76716-836BX 102.67 USD
76716-836
Lighthouse Locking Wall Mount Unit for Sharps Containers
Bins Sharps Containers
Secures sharps containers in a sturdy locked enclosure.
  • All cabinets are conveniently keyed alike
  • Mounting hardware included
  • Holds Kendall/Covidien 5 quart sharps container with counter balanced lid
  • Made from sturdy ABS plastic for extra protection of the sharps container and long-lasting use
Helps maintain OSHA compliance by keeping used sharps away from patients and personnel, and preventing tipping and spilling of contents. Includes a five quart locking wall mount unit, and a five quart Sharpstar transparent red sharps container.