Lighthouse Phlebotomy Sharps Container
Supplier: Unimed-Midwest
SR1Q100900
76716-832BX 92.93 USD
76716-832
Lighthouse Phlebotomy Sharps Container
Bins Sharps Containers
Wall mount bracket and tabletop holder available separately.
  • Adapts to present blood drawing trays and techniques
  • Facilitates disconnection of multi-sample needles from needle holders
  • Locks for final disposal"
1 quart red phlebotomy sharps disposal container.