Lighthouse Scout Electrostatic Backpack Sprayer
Supplier: Unimed-Midwest
SCOUTES200
76716-830EA 5080.45 USD
76716-830
Lighthouse Scout Electrostatic Backpack Sprayer
Spray Guns
High-efficiency touchless sprayer / disinfectant applicator and operational intelligence platform.
  • In-field service design delivers maximum uptime and reliability
  • Integrated data capture, alerts, and reporting platform drive operational intelligence and improvements
  • High-efficiency sprayer saves time and reduces chemical usage
  • Tool-free nozzle changes
  • Pump replacement in under 10 minutes with kit
The Scout ES200 delivers best-in-class efficiency, reliability, and performance to maximize cleaning and disinfecting efforts in any environment. Powered by Lighthouse Connect, Scout lets users capture, report, and leverage key disinfection data to optimize the efficiency and efficacy of infection prevention programs.
Delivery information: Supplied with:
• 2 Gallon Chemistry & Rinse Tank
• 4’ Electrostatic-Fluid Hose
• Scout ES200 Spray Wand
• 18” Spray Nozzle Extender
• 2x 75 Micron Spray Nozzles
• 2x Battery: 1 in Sprayer, 1 Spare
• Battery Charger and Cord
• Operator Grounding Wrist Strap