ThinkLite Flair 2.0 Indoor Air Quality Monitor
Supplier: Unimed-Midwest
TASM103V20
76716-826EA 4375.45 USD
76716-826
ThinkLite Flair 2.0 Indoor Air Quality Monitor
Monitoring Systems CO2 and Temperature Monitoring Systems
The ThinkLite Flair 2.0 Indoor Air Quality Monitor can help monitor and track the air quality inside a facility or environment.
  • Advanced dashboard design and functionality
  • Custom alert features for 24-hour safety support
  • Sleek new design is 25% smaller than previous models
  • Enhanced security with the ability to secure the front cover
  • PoE feature, 2 USB/C ports, plus 2 ethernet ports
  • Additional air input holes for better air monitoring accuracy
  • 5000 sqare foot coverage
  • Up to 2-hour battery life
The Flair IAQ monitor provides 24/7 continuous indoor air quality monitoring, allowing users to quickly remediate health risks. Indoor environments can have a profound impact on personal health, from productivity, mood, energy, to overall well-being. The Flair IAQ monitor provides peace of mind, opening the door for 24/7-access to data from any indoor environment.