Lighthouse Econo Emergency Spill Kit
Supplier: Unimed-Midwest
KIT-BMW
76716-802EA 24.73 USD
76716-802
Lighthouse Econo Emergency Spill Kit
Spill Control Materials
Everything needed to clean and disinfect small biohazard spills.
  • Quick, simple, and economical
  • Essential PPE included
  • Cleanup and disinfection products
  • Complies with OSHA Bloodborne Pathogen Standard
  • Easy-to-use kit is designed for non-medical personnel
  • Stores in a 9×12" clear zip bag
Kit includes: (1) Bio-Wipe Bag™, (1) 2 oz. Cavicide disinfectant spray, (1) pair of Nitrile gloves, (1) heavy-duty towelette, (1) antimicrobial hand wipe, and (1) instruction sheet, printed in English and Spanish.