Lighthouse Launderable Microfiber Wipe
Supplier: Unimed-Midwest
TMMP446MW2
76716-800CS 94.27 USD
76716-800
Lighthouse Launderable Microfiber Wipe
Wipes Reusable Cleaning Cloths Cloths for General Applications
The launderable microfiber wipe can be used both as a dry and wet wipe with varying chemistries to clean surfaces in any facility or environment.
  • Effective in removing dirt and bacteria
  • Saves money and resources
  • Eco-friendly
  • Can be washed and reused
Microfiber wipes can remove dirt and grime more effectively because the fibers attract and hold onto dirt particles. Washable microfiber wipes can be used for hundreds of cleanings before needing to be replaced, which saves money on new purchases and reduces waste.