Lighthouse Biohazard Spill Cleanup Kit
Supplier: Unimed-Midwest
KIT-B
76716-798EA 32.7 USD
76716-798
Lighthouse Biohazard Spill Cleanup Kit
Spill Control Materials
Everything needed to clean and disinfect small biohazard spills.
  • Quick, simple, and economical
  • Essential PPE included
  • Cleanup and disinfection products
  • Complies with OSHA Bloodborne Pathogen Standard
  • Easy-to-use kit is designed for non-medical personnel
  • Stores in a 9×12" clear zip bag
Kit includes: (1) red biohazard waste bag, (1) FluidShield fog-free procedure mask, (1) fluid-resistant barrier gown, (1) 20 pack of 6×7" Accel TB Surface Disinfectant wipes with a 1 minute contact time against bacteria and viruses, including Norovirus, (1) pair of Nitrile gloves, (5) heavy-duty 3-ply towels, (1) antiseptic towelette hand wipe, and (1) detailed instruction sheet with photos.