Lighthouse Complete In-Room Sharps Container System
Supplier: Unimed-Midwest
SSGB00056H
76716-760BX 73.29 USD
76716-760
Lighthouse Complete In-Room Sharps Container System
Bins Sharps Containers
Secures sharps containers in a sturdy locked enclosure.
  • All cabinets are conveniently keyed alike
  • Mounting hardware included
  • Holds Kendall/Covidien 5 quart sharps container with counter balanced lid
  • Single glove box holder slides onto bottom of unit
  • Made from sturdy ABS plastic for extra protection of the sharps container and long-lasting use
Helps maintain OSHA compliance by keeping used sharps away from patients and personnel, and preventing tipping and spilling of contents.
The Complete In-Room Sharps Container System includes a 5 quart locking wall mount unit, a five quart Sharpstar transparent red sharps container, and one glove box holder.