SYBR® Green based qPCR 2X master mix, G-Biosciences
Supplier: G-Biosciences
SYBR® Green based qPCR 2x master mix contains chemically modified hot start Taq DNA polymerase, SYBR® Green I fluorescent dye, low ROX reference dye, dNTPs and optimized buffer with stabilizers. This master mix is ideal for high-throughput real-time PCR screening and validation.
- Enhanced efficiency, specificity, and sensitivity
- Compatible with all real-time PCR instruments
- Superior gene expression results under various cycling conditions
Applications:
Gene expression analysis
Genotyping
Mutation detection, pathogen detection, viral detection, absolute quantification
Caution: Stable for one year when stored at -20 °C.