trans-Platin 98%

Supplier: Ambeed
Danger

Synonyms: trans-Diamminedichloroplatinum (II), trans-Diammineplatinum (II) dichloride, trans-Dichlorodiammine platinum(II), trans-Dichlorodiamineplatinum(II)

A993549-100MG A993549-250MG A993549-1G
76698-950EA 15 USD
76698-950 76698-952 77880-811
trans-Platin 98%
trans-Platin

Caution: for Research Use Only

Formula: H₆Cl₂N₂Pt
MDL Number: MFCD00011623
CAS Number: 14913-33-8
UN: 3288
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR