Calcon® (mordant black 17) 70+%

Supplier: Ambeed
Warning

A898143-1000G A898143-100G A898143-25G A898143-500G
76698-578EA 97 USD
76698-578 76698-580 76698-582 76698-584
Calcon® (mordant black 17) 70+%
Calcon® (Mordant Black 17)

Caution: for Research Use Only

Formula: C₂₀H₁₃N₂NaO₅S
MDL Number: MFCD00004069
CAS Number: 2538-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR