Copper(II) carbonate basic 54-57% Cu

Supplier: Ambeed
Danger

Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)

A802040-1000G A802040-100G A802040-25G A802040-500G
76698-054EA 25 USD
76698-054 76698-056 76698-058 76698-060
Copper(II) carbonate basic 54-57% Cu
Copper (II) carbonate basic

Caution: for Research Use Only

Formula: CuCO₃·Cu(OH)₂
MDL Number: MFCD00010976
CAS Number: 12069-69-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR