Cesium acetate 99%

Supplier: Ambeed
Warning

Synonyms: Acetic acid cesium salt

A743677-100G A743677-10G A743677-25G A743677-5G A743677-500G
76697-672EA 55 USD
76697-672 76697-674 76697-676 76697-678 77627-490
Cesium acetate 99%
Cesium acetate

Caution: for Research Use Only

Formula: H₃CCOOCs
MDL Number: MFCD00013056
CAS Number: 3396-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR