Cesium acetate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Acetic acid cesium salt
A743677-100G
A743677-10G
A743677-25G
A743677-5G
A743677-500G
76697-672EA
55
USD
76697-672
76697-674
76697-676
76697-678
77627-490
Cesium acetate 99%
Cesium acetate
Caution: for Research Use Only
Formula:
H₃CCOOCs |
MDL Number:
MFCD00013056 CAS Number: 3396-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...