Barium stearate 98+%

Supplier: Ambeed
Warning

Synonyms: Stearic acid barium salt

A643051-1000G A643051-100G A643051-500G
76696-910EA 20 USD
76696-910 76696-912 76696-914
Barium stearate 98+%
Barium stearate

Caution: for Research Use Only

Formula: C₃₆H₇₀BaO₄
MDL Number: MFCD00050711
CAS Number: 6865-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR