Coumarin 99+%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone
A633268-1000G
A633268-100G
A633268-100MG
A633268-10G
A633268-25G
A633268-500G
76696-822EA
49
USD
76696-822
76696-824
76696-826
76696-828
76696-830
76696-832
Coumarin 99+%
Coumarin
Caution: for Research Use Only
Formula:
C₉H₆O₂ |
MDL Number:
MFCD00006850 CAS Number: 91-64-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...