Acetylacetonato(1,5-cyclooctadiene)rhodium(I) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)
A567452-1G
A567452-250MG
76696-266EA
483
USD
76696-266
76696-268
Acetylacetonato(1,5-cyclooctadiene)rhodium(I) 98%
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
Caution: for Research Use Only
Formula:
C₁₃H₂₀O₂Rh |
MDL Number:
MFCD00075046 CAS Number: 12245-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...