Calcium acetate 99%

Supplier: Ambeed
Warning

A466775-100G A466775-25G A466775-500G A466775-1000G
76695-418EA 7 USD
76695-418 76695-420 76695-422 77594-470
Calcium acetate 99%
Calcium acetate

Caution: for Research Use Only

Formula: (H₃CCOO)₂Ca
MDL Number: MFCD00150019
CAS Number: 62-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR