Calcium acetate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Calcium acetate, United States Pharmacopeia (USP) Reference Standard, Calcium di(acetate), Acetic acid calcium salt
A466775-100G
A466775-25G
A466775-500G
A466775-1000G
76695-418EA
7
USD
76695-418
76695-420
76695-422
77594-470
Calcium acetate 99%
Calcium acetate
Caution: for Research Use Only
Formula:
(H₃CCOO)₂Ca MW: 158.17 g/mol |
MDL Number:
MFCD00150019 CAS Number: 62-54-4 Merck Index: 12,01683 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...