Carbonylhydridotris(triphenylphosphine)rhodium(I) 98%

Supplier: Ambeed
Danger

Synonyms: Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)

A441776-1G A441776-250MG A441776-5G A441776-100MG
76695-154EA 120 USD
76695-154 76695-156 76695-158 77601-914
Carbonylhydridotris(triphenylphosphine)rhodium(I) 98%
Carbonylhydridotris(triphenylphosphine)rhodium(I)

Caution: for Research Use Only

Formula: C₅₅H₄₆OP₃Rh
MW: 918.80 g/mol
MDL Number: MFCD00151644
CAS Number: 17185-29-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR