(1-Butyl)triphenylphosphonium bromide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Butyltriphenylphosphonium bromide
A427714-1000G
A427714-100G
A427714-500G
A427714-10G
A427714-25G
76695-036EA
59
USD
76695-036
76695-038
76695-040
77601-836
77626-854
(1-Butyl)triphenylphosphonium bromide 97%
(1-Butyl)triphenylphosphonium bromide
Caution: for Research Use Only
Formula:
H₃C(CH₂)₃P(C₆H₅)₃Br |
MDL Number:
MFCD00011855 CAS Number: 1779-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...