7-Methoxy-4-(trifluoromethyl)coumarin 97%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-4-(trifluoromethyl)-2H-chromen-2-one, Methyl 4-(trifluoromethyl)umbelliferyl ether

A426846-1G A426846-250MG
76695-032EA 75 USD
76695-032 76695-034
7-Methoxy-4-(trifluoromethyl)coumarin 97%
7-Methoxy-4-(trifluoromethyl)coumarin

Caution: for Research Use Only

Formula: C₁₁H₇F₃O₃
MDL Number: MFCD00132980
CAS Number: 575-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR