Benzyl acrylate 97% stabilised

Supplier: Ambeed
Warning

Synonyms: Acrylic Acid Benzyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A386639-100G A386639-10G A386639-1G A386639-25G A386639-5G A386639-1000G A386639-500G
76694-582EA 22 USD
76694-582 76694-584 76694-586 76694-588 76694-590 77572-152 77594-176
Benzyl acrylate 97% stabilised
Benzyl acrylate

Caution: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00048147
CAS Number: 2495-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR