Decanenitrile 98%

Supplier: Ambeed
Warning

A335741-25G A335741-5G
76693-940EA 30 USD
76693-940 77879-721
Decanenitrile 98%
Decanenitrile

Caution: for Research Use Only

Formula: C₁₀H₁₉N
MDL Number: MFCD00039505
CAS Number: 1975-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR