Diammonium tetrachloropalladate Pd 36% min
Supplier: Ambeed
|
Warning
|
Synonyms:
Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)
A309867-1G
A309867-250MG
A309867-5G
76693-544EA
48
USD
76693-544
76693-546
77626-546
Diammonium tetrachloropalladate Pd 36% min
Diammonium tetrachloropalladate
Caution: for Research Use Only
Formula:
H₈Cl₄N₂Pd |
MDL Number:
MFCD00010884 CAS Number: 13820-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...