Chalcone 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Diphenyl-2-propen-1-one, Phenyl Styryl ketone, 2-Benzalacetophenone, 2-Benzylideneacetophenone

A256328-100G A256328-25G A256328-500G A256328-5G
76692-628EA 42 USD
76692-628 76692-630 76692-632 76692-634
Chalcone 98%
Chalcone

Caution: for Research Use Only

Formula: (C₆H₅)C(O)CHCH(C₆H₅)
MW: 208.26 g/mol
Boiling Pt: 345
MDL Number: MFCD00003082
CAS Number: 94-41-7
Merck Index: 13,02045

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR