Chalcone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Diphenyl-2-propen-1-one, Phenyl Styryl ketone, 2-Benzalacetophenone, 2-Benzylideneacetophenone
A256328-100G
A256328-25G
A256328-500G
A256328-5G
76692-628EA
42
USD
76692-628
76692-630
76692-632
76692-634
Chalcone 98%
Chalcone
Caution: for Research Use Only
Formula:
(C₆H₅)C(O)CHCH(C₆H₅) MW: 208.26 g/mol Boiling Pt: 345 |
MDL Number:
MFCD00003082 CAS Number: 94-41-7 Merck Index: 13,02045 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...