Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1,5-Cyclooctadiene)rhodium chloride dimer
A254450-100MG
A254450-250MG
A254450-1G
A254450-5G
103812-140EA
290
USD
103812-140
103851-994
76692-588
76692-590
77091-436
77091-438
Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%
Chloro(1,5-cyclooctadiene)rhodium (I) dimer
Caution: for Research Use Only
Formula:
C₁₆H₂₄Cl₂Rh₂ MW: 493.08 g/mol Melting Pt: 243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012415 CAS Number: 12092-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...