(4-Fluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Fluorophenyl)acetic acid

A242168-100G A242168-10G A242168-25G A242168-500G A242168-1000G
76692-308EA 33 USD
76692-308 76692-310 76692-312 76692-314 77877-806
(4-Fluorophenyl)acetic acid 98%
(4-Fluorophenyl)acetic acid

Caution: for Research Use Only

Formula: (C₆H₄F)CH₂COOH
MDL Number: MFCD00004343
CAS Number: 405-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR